Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCNY SIRNA Assays ›
Gene Symbol:
CCNY
Gene Name:
cyclin Y
Gene Aliases:
C10orf9, CBCP1, CCNX, CFP1
Chromosome Location:
Chr.10: 35246981 - 35572670 on Build GRCh38
UniGene ID:
Hs.14745
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
129281
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCNY
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282852.1
NP_001269781.1
6
620
NM_001282853.1
NP_001269782.1
8
758
NM_001282854.1
NP_001269783.1
5
558
NM_145012.5
NP_659449.3
7
695
NM_181698.3
NP_859049.2
9
719
XM_006717392.3
XP_006717455.1
7
464
XM_011519357.2
XP_011517659.1
6
654
XM_011519358.1
XP_011517660.1
10
786
XM_011519360.2
XP_011517662.1
11
888
XM_011519361.1
XP_011517663.1
11
770
XM_011519362.1
XP_011517664.1
10
750
XM_011519363.1
XP_011517665.1
10
763
XM_011519364.1
XP_011517666.1
12
884
XM_011519365.2
XP_011517667.1
11
803
XM_011519366.1
XP_011517668.1
10
733
XM_011519367.1
XP_011517669.1
8
563
XM_011519368.2
XP_011517670.1
7
458
XM_011519369.2
XP_011517671.1
6
948
XM_017015839.1
XP_016871328.1
7
381
XM_017015840.1
XP_016871329.1
7
383
XM_017015841.1
XP_016871330.1
7
381
GenBank mRNA
AF429969.1
-
9
675
AF465728.1
-
7
441
AF465729.1
-
7
516
AK057280.1
-
9
675
AK303283.1
-
5
412
AK314666.1
-
8
734
AL834355.1
-
7
553
BC069224.1
-
7
521
BC094815.1
-
7
547
6340750
BC104773.1
-
8
504
8143776
BC104801.1
-
8
504
8143804
BC143450.1
-
7
397
9051958
BC143455.1
-
8
504
9051963
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001191833.
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
Wnt signaling pathway
positive regulation of cyclin-dependent protein serine/threonine kinase activity
cell division
regulation of canonical Wnt signaling pathway
protein binding
cyclin-dependent protein serine/threonine kinase regulator activity
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0