Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIGD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145719.2 | NP_663771.1 | 2 | 1382 | ||
GenBank mRNA | AF001546.1 | - | 1 | 548 | ||
BC074862.2 | - | 1 | 1240 | 30915228 |