Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIGD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145720.3 | NP_663772.1 | 2 | 2251 | ||
XM_005262807.4 | XP_005262864.1 | 3 | 2354 | |||
GenBank mRNA | AK058054.1 | - | 1 | 1221 | ||
BC037869.2 | - | 2 | 2228 | 5273281 |