Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282506.1 | NP_001269435.1 | 2, 3 | 2093 | ||
NM_001291833.1 | NP_001278762.1 | 3, 4 | 1568 | |||
NM_152673.3 | NP_689886.3 | 3, 4 | 1580 | |||
GenBank mRNA | AB098731.1 | - | 2, 3 | 1831 | ||
AK027314.1 | - | 4, 5 | 1331 | |||
AK075138.1 | - | 3, 4 | 1289 | |||
AK311504.1 | - | 3, 4 | 1578 | |||
AY358449.1 | - | 3, 4 | 1512 | |||
BC044243.1 | - | 3, 4 | 1467 | 5754328 | ||
BF883124.1 | - | 1, 2 | 400 |