Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152721.5 | NP_689934.2 | 2 | 356 | ||
XM_011525875.2 | XP_011524177.1 | 2 | 377 | |||
GenBank mRNA | AK057795.1 | - | 1 | 65 | ||
AY599248.1 | - | 2 | 354 | |||
BC096744.1 | - | 2 | 349 | 5260167 | ||
DB480182.1 | - | 2 | 356 |