Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193341.1 | NP_001180270.1 | 1 | 185 | ||
NM_152732.4 | NP_689945.2 | 1 | 185 | |||
XM_005248901.3 | XP_005248958.1 | 1 | 158 | |||
XM_006715014.1 | XP_006715077.1 | 1 | 185 | |||
XM_017010426.1 | XP_016865915.1 | 1 | 158 | |||
GenBank mRNA | AK055407.1 | - | 1 | 125 | ||
AK291399.1 | - | 1 | 158 | |||
BC029519.1 | - | 1 | 142 | 5296237 | ||
DA486918.1 | - | 1 | 186 |