Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BEND7 SIRNA Assays ›
Gene Symbol:
BEND7
Gene Name:
BEN domain containing 7
Gene Aliases:
C10orf30
Chromosome Location:
Chr.10: 13438484 - 13528772 on Build GRCh38
UniGene ID:
Hs.498740
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
129564
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BEND7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001100912.1
NP_001094382.1
2
246
NM_152751.2
NP_689964.2
3
416
XM_005252402.4
XP_005252459.1
3
514
XM_011519390.2
XP_011517692.1
2
168
XM_011519391.2
XP_011517693.1
2
168
XM_011519392.2
XP_011517694.1
3
513
XM_011519393.2
XP_011517695.1
2
263
XM_011519394.2
XP_011517696.1
4
410
XM_011519395.2
XP_011517697.1
2
770
XM_011519396.2
XP_011517698.1
3
861
XM_011519397.2
XP_011517699.1
3
416
XM_011519398.2
XP_011517700.1
2
168
XM_011519399.2
XP_011517701.1
2
168
XM_011519400.2
XP_011517702.1
2
168
XM_011519401.2
XP_011517703.1
2
168
XM_011519402.2
XP_011517704.1
2
168
XM_011519403.2
XP_011517705.1
2
168
XM_011519404.2
XP_011517706.1
2
168
XM_011519405.2
XP_011517707.1
2
168
XM_011519406.2
XP_011517708.1
2
168
XM_011519407.1
XP_011517709.1
2
168
XM_011519408.2
XP_011517710.1
2
168
XM_011519410.2
XP_011517712.1
2
168
XM_011519411.2
XP_011517713.1
2
168
XM_011519412.1
XP_011517714.1
2
168
XM_011519413.1
XP_011517715.1
2
168
XM_011519414.1
XP_011517716.1
2
168
XM_017015915.1
XP_016871404.1
4
395
XM_017015916.1
XP_016871405.1
2
168
XM_017015917.1
XP_016871406.1
2
168
XM_017015918.1
XP_016871407.1
3
448
XM_017015919.1
XP_016871408.1
3
470
GenBank mRNA
AK097602.1
-
3
416
BC031618.1
-
2
215
5172600
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0