Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCSAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190259.1 | NP_001177188.1 | 6 | 864 | ||
NM_001190260.1 | NP_001177189.1 | 5 | 813 | |||
NM_152785.4 | NP_689998.1 | 6 | 858 | |||
XM_005247272.2 | XP_005247329.1 | 6 | 1282 | |||
XM_011512627.2 | XP_011510929.1 | 6 | 736 | |||
XM_011512628.2 | XP_011510930.1 | 6 | 793 | |||
XM_011512629.1 | XP_011510931.1 | 6 | 1285 | |||
XM_011512630.1 | XP_011510932.1 | 6 | 1291 | |||
GenBank mRNA | AF521911.1 | - | 6 | 845 | ||
AY212246.1 | - | 6 | 779 | |||
BC030506.1 | - | 6 | 726 | 4385178 |