Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCSAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190259.1 | NP_001177188.1 | 6 | 2048 | ||
NM_001190260.1 | NP_001177189.1 | 5 | 1997 | |||
NM_152785.4 | NP_689998.1 | 6 | 2042 | |||
XM_005247272.2 | XP_005247329.1 | 6 | 2466 | |||
XM_011512627.2 | XP_011510929.1 | 6 | 1920 | |||
XM_011512628.2 | XP_011510930.1 | 6 | 1977 | |||
XM_011512629.1 | XP_011510931.1 | 6 | 2469 | |||
XM_011512630.1 | XP_011510932.1 | 6 | 2475 | |||
GenBank mRNA | AY212246.1 | - | 6 | 1961 |