Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153033.4 | NP_694578.1 | 4 | 1665 | ||
GenBank mRNA | AK055201.1 | - | 2 | 1004 | ||
AK056631.1 | - | 4 | 1588 |