Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017696.2 | NP_060166.2 | 2 | 506 | ||
NM_153255.4 | NP_694987.1 | 3 | 778 | |||
GenBank mRNA | AK122665.1 | - | 3 | 618 | ||
BC031658.1 | - | 3 | 730 | 5177145 | ||
BX649114.1 | - | 1 | 112 | |||
KU178895.1 | - | 2 | 491 | |||
KU178896.1 | - | 3 | 582 |