Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173605.1 | NP_775876.1 | 2 | 1156 | ||
NM_199464.2 | NP_955751.1 | 3 | 1254 | |||
GenBank mRNA | AF086244.1 | - | 1 | 122 | ||
AY129653.1 | - | 3 | 1064 | |||
AY129654.1 | - | 2 | 974 | |||
AY169387.1 | - | 3 | 1254 | |||
AY169388.1 | - | 2 | 1156 | |||
BC020887.1 | - | 3 | 1064 | 4690662 | ||
BC032343.1 | - | 2 | 950 | 5207702 | ||
BX116021.1 | - | 1 | 117 |