Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP11C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010986.2 | NP_001010986.1 | 9 | 861 | ||
NM_173694.4 | NP_775965.2 | 9 | 861 | |||
XM_017029439.1 | XP_016884928.1 | 10 | 867 | |||
XM_017029440.1 | XP_016884929.1 | 10 | 1538 | |||
XM_017029441.1 | XP_016884930.1 | 11 | 947 | |||
XM_017029442.1 | XP_016884931.1 | 10 | 1088 | |||
XM_017029443.1 | XP_016884932.1 | 10 | 866 | |||
XM_017029444.1 | XP_016884933.1 | 9 | 1478 | |||
XM_017029445.1 | XP_016884934.1 | 10 | 868 | |||
XM_017029446.1 | XP_016884935.1 | 10 | 870 | |||
XM_017029447.1 | XP_016884936.1 | 10 | 1071 | |||
XM_017029448.1 | XP_016884937.1 | 10 | 1203 | |||
XM_017029449.1 | XP_016884938.1 | 9 | 809 | |||
GenBank mRNA | AJ580093.1 | - | 9 | 863 | ||
AJ580094.1 | - | 9 | 863 | |||
BC132728.1 | - | 9 | 799 | |||
BC132730.1 | - | 9 | 799 |