Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003334.3 | NP_003325.2 | 4 | 522 | ||
NM_153280.2 | NP_695012.1 | 4 | 450 | |||
XM_005272649.1 | XP_005272706.1 | 5 | 900 | |||
XM_011543954.2 | XP_011542256.1 | 5 | 542 | |||
XM_017029777.1 | XP_016885266.1 | 4 | 1617 | |||
XM_017029778.1 | XP_016885267.1 | 5 | 2791 | |||
XM_017029779.1 | XP_016885268.1 | 4 | 3299 | |||
XM_017029780.1 | XP_016885269.1 | 4 | 407 | |||
XM_017029781.1 | XP_016885270.1 | 4 | 455 | |||
GenBank mRNA | BC013041.2 | - | 4 | 389 | 3543021 | |
BC020261.1 | - | 1 | 84 | |||
BI547207.1 | - | 4 | 528 | |||
M58028.1 | - | 4 | 428 | |||
X56976.1 | - | 4 | 331 |