Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCSK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174936.3 | NP_777596.2 | 12 | 3313 | ||
NR_110451.1 | 10 | 2558 | ||||
GenBank mRNA | AK075365.1 | - | 12 | 3241 | ||
AK122717.1 | - | 13 | 2998 | |||
AK124635.1 | - | 8 | 3497 |