Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDYE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175064.2 | NP_778234.2 | 7 | 2243 | ||
XM_005249719.3 | XP_005249776.1 | 9 | 2754 | |||
XM_017012015.1 | XP_016867504.1 | 9 | 2692 | |||
XM_017012016.1 | XP_016867505.1 | 8 | 2104 | |||
GenBank mRNA | AF412027.1 | - | 7 | 2254 | ||
AL137266.1 | - | 7 | 2027 |