Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024647.3 | NP_001019818.1 | 3 | 359 | ||
NM_001278402.1 | NP_001265331.1 | 3 | 274 | |||
NM_022456.4 | NP_071901.2 | 5 | 1010 | |||
NM_175623.3 | NP_783322.1 | 5 | 1144 | |||
NM_175624.3 | NP_783323.1 | 5 | 1010 | |||
NM_175625.3 | NP_783324.1 | 5 | 1144 | |||
NR_103519.1 | 5 | 1010 | ||||
NR_103520.1 | 5 | 1144 | ||||
XM_006719226.3 | XP_006719289.3 | 5 | 1133 | |||
XM_006719227.3 | XP_006719290.3 | 5 | 806 | |||
XM_017018773.1 | XP_016874262.1 | 5 | 768 | |||
XM_017018774.1 | XP_016874263.1 | 7 | 1021 | |||
XM_017018776.1 | XP_016874265.1 | 6 | 893 | |||
XM_017018777.1 | XP_016874266.1 | 6 | 1312 | |||
GenBank mRNA | AJ312896.1 | - | 5 | 834 | ||
AJ312897.1 | - | 5 | 1126 | |||
AJ312898.1 | - | 5 | 834 | |||
AJ312899.1 | - | 5 | 1126 | |||
AJ312900.1 | - | 5 | 834 | |||
AJ312901.1 | - | 5 | 1126 | |||
AK026201.1 | - | 5 | 834 | |||
AK027566.1 | - | 3 | 270 | |||
AK027671.1 | - | 3 | 260 | |||
AK057844.1 | - | 5 | 739 | |||
AK225441.1 | - | 5 | 834 | |||
AK314547.1 | - | 5 | 845 | |||
BC015548.1 | - | 3 | 337 | 3846338 | ||
BC059358.1 | - | 5 | 799 | 5300636 | ||
DN912041.1 | - | 3 | 359 |