Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1S3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039569.1 | NP_001034658.1 | 5 | 1309 | ||
NR_110905.1 | 6 | 1348 | ||||
NR_110906.1 | 4 | 1200 | ||||
XM_011510600.2 | XP_011508902.1 | 4 | 1151 | |||
GenBank mRNA | AK025338.1 | - | 4 | 1168 | ||
BC021898.1 | - | 6 | 1292 | 4340257 |