Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEZ6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098635.1 | NP_001092105.1 | 16 | 3106 | ||
NM_001290202.1 | NP_001277131.1 | 16 | 2942 | |||
NM_178860.4 | NP_849191.3 | 16 | 3106 | |||
GenBank mRNA | AF502129.1 | - | 16 | 3194 | ||
AF502130.1 | - | 8 | 1791 | |||
AK054913.1 | - | 7 | 967 | |||
AK055383.1 | - | 10 | 1378 | |||
AK091522.1 | - | 16 | 2942 | |||
AK096473.1 | - | 15 | 2355 | |||
AK125377.1 | - | 16 | 3067 | |||
AK223620.1 | - | 16 | 3068 | |||
AL834405.1 | - | 15 | 2851 | |||
AY038048.1 | - | 16 | 3097 |