Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271856.1 | NP_001258785.1 | 7 | 1670 | ||
NM_181711.3 | NP_859062.1 | 8 | 1904 | |||
XM_005268691.3 | XP_005268748.1 | 8 | 3072 | |||
XM_011537996.1 | XP_011536298.1 | 7 | 2850 | |||
GenBank mRNA | AJ227899.1 | - | 1 | 608 | ||
BC035500.2 | - | 7 | 1641 | 4422992 | ||
BC052967.1 | - | 7 | 1588 | |||
BC053666.1 | - | 8 | 1885 | 6151168 |