Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC36A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145017.1 | NP_001138489.1 | 11 | 2269 | ||
NM_181774.3 | NP_861439.3 | 10 | 2146 | |||
XM_011537627.2 | XP_011535929.1 | 10 | 2068 | |||
XM_011537630.1 | XP_011535932.1 | 8 | 1493 | |||
XM_011537631.1 | XP_011535933.1 | 8 | 1504 | |||
XM_011537633.1 | XP_011535935.1 | 6 | 1356 | |||
GenBank mRNA | AK127978.1 | - | 10 | 2139 | ||
AY162215.1 | - | 10 | 1964 |