Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085357.1 | NP_001078826.1 | 2 | 362 | ||
NM_181780.3 | NP_861445.3 | 2 | 362 | |||
XM_011512447.2 | XP_011510749.1 | 3 | 456 | |||
XM_017005748.1 | XP_016861237.1 | 3 | 437 | |||
GenBank mRNA | AI792952.1 | - | 2 | 330 | ||
AJ717664.1 | - | 2 | 159 | |||
AK131204.1 | - | 2 | 151 | |||
AK292692.1 | - | 2 | 341 | |||
AY293286.1 | - | 2 | 159 | |||
AY599411.1 | - | 2 | 159 | |||
AY606805.1 | - | 2 | 159 | |||
BC107091.1 | - | 2 | 159 | 40015588 | ||
BC107092.1 | - | 2 | 157 | |||
CD702861.1 | - | 2 | 400 | |||
DQ198368.1 | - | 2 | 159 |