Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCOA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122842.2 | NP_001116314.1 | 3 | 530 | ||
NM_001199619.1 | NP_001186548.1 | 4 | 513 | |||
NM_001199620.1 | NP_001186549.1 | 5 | 631 | |||
NM_181782.4 | NP_861447.3 | 3 | 530 | |||
XM_005266822.4 | XP_005266879.1 | 3 | 560 | |||
XM_006715340.3 | XP_006715403.1 | 3 | 487 | |||
XM_011535455.2 | XP_011533757.1 | 3 | 435 | |||
XM_017010269.1 | XP_016865758.1 | 3 | 246 | |||
XM_017010270.1 | XP_016865759.1 | 4 | 492 | |||
XM_017010271.1 | XP_016865760.1 | 3 | 490 | |||
XM_017010272.1 | XP_016865761.1 | 3 | 343 | |||
XM_017010273.1 | XP_016865762.1 | 3 | 560 | |||
XM_017010274.1 | XP_016865763.1 | 3 | 649 | |||
GenBank mRNA | AB074157.1 | - | 3 | 330 | ||
AF493978.1 | - | 4 | 513 | |||
AK127512.1 | - | 3 | 340 | |||
AK307395.1 | - | 4 | 492 | |||
AL832621.2 | - | 3 | 323 | |||
AL832628.2 | - | 3 | 287 | |||
BC036461.1 | - | 3 | 358 | |||
BC071782.1 | - | 2 | 298 | |||
BC137094.1 | - | 5 | 547 | 9021091 | ||
BC137095.1 | - | 4 | 429 | 9021092 | ||
BX537385.1 | - | 3 | 325 | |||
DA770225.1 | - | 3 | 530 |