Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181843.2 | NP_862826.1 | 3 | 808 | ||
GenBank mRNA | BC018644.1 | - | 3 | 801 | 3956297 |