Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182832.2 | NP_878252.2 | 1 | 424 | ||
GenBank mRNA | AK092431.1 | - | 1 | 424 | ||
BC093685.1 | - | 1 | 369 | 7939530 | ||
BC101615.1 | - | 1 | 369 | 8069121 | ||
BC101617.1 | - | 1 | 369 | 8069123 |