Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039753.2 | NP_001034842.2 | 12, 13 | 1990 | ||
XM_017004098.1 | XP_016859587.1 | 12, 13 | 2143 | |||
XM_017004099.1 | XP_016859588.1 | 12, 13 | 2143 | |||
XM_017004100.1 | XP_016859589.1 | 12, 13 | 2145 | |||
XM_017004101.1 | XP_016859590.1 | 6, 7 | 977 | |||
XM_017004102.1 | XP_016859591.1 | 13, 14 | 2764 | |||
GenBank mRNA | BX647078.1 | - | 2, 3 | 287 |