Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152696.3 | NP_689909.2 | 12 | 2572 | ||
NM_181358.2 | NP_852003.1 | 10 | 1292 | |||
NM_198268.2 | NP_938009.1 | 12 | 2572 | |||
NM_198269.2 | NP_938010.1 | 11 | 1483 | |||
XM_005270609.3 | XP_005270666.1 | 12 | 2749 | |||
XM_005270610.4 | XP_005270667.1 | 12 | 2746 | |||
XM_005270611.4 | XP_005270668.1 | 12 | 2749 | |||
XM_005270612.4 | XP_005270669.1 | 12 | 2711 | |||
XM_005270613.4 | XP_005270670.1 | 14 | 2662 | |||
XM_006710443.3 | XP_006710506.1 | 12 | 2647 | |||
XM_011540977.2 | XP_011539279.1 | 11 | 2629 | |||
XM_017000605.1 | XP_016856094.1 | 12 | 2708 | |||
GenBank mRNA | AB014530.1 | - | 3 | 252 | ||
AB089957.1 | - | 12 | 2536 | |||
AK122662.1 | - | 12 | 2570 | |||
AK302726.1 | - | 12 | 2756 | |||
AL833829.1 | - | 11 | 1461 | |||
BC028408.1 | - | 11 | 1488 | 4821915 | ||
BC033012.2 | - | 12 | 2572 | 4830408 | ||
BC036057.1 | - | 10 | 1292 | 5272816 |