Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HPS4 SIRNA Assays ›
Gene Symbol:
HPS4
Gene Name:
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases:
BLOC3S2, LE
Chromosome Location:
Chr.22: 26443613 - 26483863 on Build GRCh38
UniGene ID:
Hs.474436
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
130263
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HPS4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022081.5
NP_071364.4
10
1439
NM_152841.2
NP_690054.1
8
1056
NR_073135.1
10
1457
NR_073136.1
9
1219
XM_006724353.3
XP_006724416.1
11
1484
XM_006724354.3
XP_006724417.1
11
1389
XM_006724360.3
XP_006724423.1
6
667
XM_011530485.1
XP_011528787.1
12
1562
XM_011530486.2
XP_011528788.1
12
1467
XM_011530487.2
XP_011528789.1
13
1672
XM_011530488.2
XP_011528790.1
12
1470
XM_011530489.2
XP_011528791.1
13
1573
XM_011530490.2
XP_011528792.1
11
1508
XM_011530491.2
XP_011528793.1
12
1562
XM_011530492.1
XP_011528794.1
12
1562
XM_011530493.2
XP_011528795.1
12
1562
XM_011530494.2
XP_011528796.1
10
858
XM_011530495.2
XP_011528797.1
4
364
XM_011530496.2
XP_011528798.1
5
199
XM_017029042.1
XP_016884531.1
10
1335
XM_017029043.1
XP_016884532.1
11
1540
XM_017029044.1
XP_016884533.1
10
1338
XM_017029045.1
XP_016884534.1
11
1508
XM_017029046.1
XP_016884535.1
10
1430
XM_017029047.1
XP_016884536.1
11
1508
XM_017029048.1
XP_016884537.1
10
1430
XM_017029049.1
XP_016884538.1
10
1335
XM_017029050.1
XP_016884539.1
10
1430
XM_017029051.1
XP_016884540.1
10
1335
XM_017029052.1
XP_016884541.1
11
1593
XM_017029053.1
XP_016884542.1
3
952
XM_017029054.1
XP_016884543.1
10
1539
XM_017029055.1
XP_016884544.1
10
1393
XM_017029056.1
XP_016884545.1
11
1606
XM_017029057.1
XP_016884546.1
11
1455
XM_017029058.1
XP_016884547.1
11
1489
XM_017029059.1
XP_016884548.1
11
1364
XM_017029060.1
XP_016884549.1
11
1363
XM_017029061.1
XP_016884550.1
11
1564
XM_017029062.1
XP_016884551.1
9
1339
XM_017029063.1
XP_016884552.1
11
1473
XM_017029064.1
XP_016884553.1
9
1222
GenBank mRNA
AB051454.1
-
9
3629
AK054717.1
-
7
1537
AK057648.1
-
11
1322
AK123691.1
-
5
1166
AK290211.1
-
10
1359
AL365512.1
-
8
960
AL713795.1
-
10
1388
AY043416.1
-
9
959
BC028625.1
-
10
1453
BC050285.1
-
10
1430
BC065030.1
-
9
1161
6148557
CR456392.1
-
9
860
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein targeting
lysosome organization
blood coagulation
hemostasis
melanocyte differentiation
positive regulation of eye pigmentation
protein stabilization
positive regulation of protein targeting to mitochondrion
protein binding
protein homodimerization activity
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0