Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POPDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308333.1 | NP_001295262.1 | 5 | 1510 | ||
NM_022135.3 | NP_071418.2 | 4 | 1427 | |||
XM_005247698.3 | XP_005247755.1 | 4 | 1568 | |||
GenBank mRNA | AF204173.1 | - | 4 | 1416 | ||
AK124602.1 | - | 1 | 4661 | |||
AK226022.1 | - | 6 | 1716 | |||
AK377174.1 | - | 7 | 907 | |||
AK377176.1 | - | 7 | 1185 | |||
AK377177.1 | - | 5 | 1189 | |||
AK377178.1 | - | 7 | 1243 | |||
AK377179.1 | - | 6 | 1275 | |||
AK377180.1 | - | 8 | 1316 | |||
AK377181.1 | - | 7 | 1324 | |||
AK377182.1 | - | 7 | 1384 | |||
AK377183.1 | - | 8 | 1445 | |||
AK377184.1 | - | 8 | 1457 | |||
BC026911.2 | - | 6 | 1057 |