Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARVG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137605.2 | NP_001131077.1 | 7, 8 | 921 | ||
NM_022141.6 | NP_071424.1 | 7, 8 | 981 | |||
XM_005261702.3 | XP_005261759.1 | 7, 8 | 708 | |||
XM_011530302.2 | XP_011528604.1 | 6, 7 | 3473 | |||
XM_017028907.1 | XP_016884396.1 | 7, 8 | 1919 | |||
XM_017028908.1 | XP_016884397.1 | 7, 8 | 1939 | |||
GenBank mRNA | AF237772.1 | - | 7, 8 | 775 | ||
AK225929.1 | - | 8, 9 | 871 | |||
AL355092.1 | - | 6, 7 | 688 | |||
AL590887.1 | - | 5, 6 | 497 | |||
BC034406.1 | - | 7, 8 | 902 | 5178529 | ||
BI818662.1 | - | 730 | ||||
BX648942.1 | - | 2, 3 | 3838 | |||
CR456480.2 | - | 6, 7 | 552 |