Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDDM3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022360.4 | NP_071755.1 | 2 | 337 | ||
GenBank mRNA | AY358865.1 | - | 2 | 337 | ||
BC128030.1 | - | 2 | 300 | 40002822 | ||
BX112699.1 | - | 2 | 333 | |||
X76386.2 | - | 1 | 299 |