Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105079.2 | NP_001098549.2 | 14 | 1898 | ||
XM_011545916.2 | XP_011544218.1 | 13 | 3245 | |||
XM_011545917.2 | XP_011544219.1 | 13 | 3245 | |||
XM_011545919.2 | XP_011544221.1 | 12 | 3208 | |||
GenBank mRNA | AK021680.1 | - | 2 | 89 | ||
AK022551.1 | - | 5 | 303 | |||
AK096322.1 | - | 6 | 681 | |||
AK298229.1 | - | 7 | 461 | |||
BQ276414.1 | - | 7 | 466 | |||
DA527487.1 | - | 6 | 322 | |||
DR761499.1 | - | 7 | 395 |