Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIGYF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022574.4 | NP_072096.2 | 8 | 1753 | ||
XM_005250532.1 | XP_005250589.1 | 10 | 1534 | |||
XM_011516472.1 | XP_011514774.1 | 10 | 1534 | |||
XM_011516477.1 | XP_011514779.1 | 10 | 1534 | |||
XM_017012526.1 | XP_016868015.1 | 10 | 1534 | |||
XM_017012527.1 | XP_016868016.1 | 12 | 1432 | |||
XM_017012528.1 | XP_016868017.1 | 11 | 1044 | |||
XM_017012529.1 | XP_016868018.1 | 11 | 1051 | |||
XM_017012530.1 | XP_016868019.1 | 11 | 1305 | |||
XM_017012531.1 | XP_016868020.1 | 6 | 557 | |||
GenBank mRNA | AY176044.1 | - | 8 | 1954 |