Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282949.2 | NP_001269878.1 | 6 | 1455 | ||
NM_001282950.2 | NP_001269879.1 | 8 | 1629 | |||
NM_001282951.2 | NP_001269880.1 | 5 | 1468 | |||
NM_001282952.2 | NP_001269881.1 | 6 | 1500 | |||
NM_001308381.1 | NP_001295310.1 | 8 | 1644 | |||
NM_022730.3 | NP_073567.1 | 7 | 1576 | |||
XM_006712693.1 | XP_006712756.1 | 8 | 1701 | |||
XM_011511638.1 | XP_011509940.1 | 7 | 3084 | |||
XM_011511639.1 | XP_011509941.1 | 7 | 1517 | |||
XM_011511640.1 | XP_011509942.1 | 7 | 1602 | |||
XM_017004716.1 | XP_016860205.1 | 7 | 1633 | |||
XM_017004717.1 | XP_016860206.1 | 6 | 1534 | |||
GenBank mRNA | AK022674.1 | - | 7 | 1507 | ||
AK024273.1 | - | 6 | 1449 | |||
AK098334.1 | - | 4 | 1913 | |||
AK124133.1 | - | 7 | 1575 | |||
AK126326.1 | - | 7 | 2831 | |||
AK222879.1 | - | 7 | 1514 | |||
BC010739.1 | - | 4 | 1111 | |||
BC091493.1 | - | 8 | 1564 | 30387712 |