Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022748.11 | NP_073585.8 | 15, 16 | 1285 | ||
XM_011515476.2 | XP_011513778.1 | 15, 16 | 2166 | |||
XM_011515477.2 | XP_011513779.1 | 15, 16 | 1318 | |||
XM_011515478.2 | XP_011513780.1 | 15, 16 | 1394 | |||
XM_011515479.2 | XP_011513781.1 | 12, 13 | 3470 | |||
XM_011515480.2 | XP_011513782.1 | 15, 16 | 1416 | |||
XM_011515481.2 | XP_011513783.1 | 13, 14 | 1082 | |||
XM_011515482.2 | XP_011513784.1 | 15, 16 | 2771 | |||
XM_011515483.2 | XP_011513785.1 | 12, 13 | 1315 | |||
XM_017012537.1 | XP_016868026.1 | 14, 15 | 1255 | |||
XM_017012538.1 | XP_016868027.1 | 15, 16 | 1562 | |||
XM_017012539.1 | XP_016868028.1 | 14, 15 | 2668 | |||
GenBank mRNA | AB208995.1 | - | 15, 16 | 1323 | ||
AF378756.1 | - | 3, 4 | 299 | |||
AF417489.1 | - | 10, 11 | 936 | |||
AK092864.1 | - | 15, 16 | 1285 | |||
AK310472.1 | - | 15, 16 | 1394 | |||
BC137133.1 | - | 11, 12 | 965 | 9021130 | ||
BC137134.1 | - | 11, 12 | 965 | 9021131 | ||
CR749644.1 | - | 15, 16 | 1234 |