Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297778.1 | NP_001284707.1 | 5 | 996 | ||
NM_022787.3 | NP_073624.2 | 5 | 981 | |||
XM_017002107.1 | XP_016857596.1 | 5 | 1134 | |||
GenBank mRNA | AF312734.1 | - | 4 | 925 | ||
AF459819.1 | - | 4 | 893 | |||
AK026065.1 | - | 5 | 959 | |||
AK222920.1 | - | 5 | 959 | |||
BC014943.1 | - | 5 | 980 | 4874147 |