Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022901.2 | NP_075052.1 | 5 | 1794 | ||
GenBank mRNA | AK024955.1 | - | 5 | 1774 | ||
AK025070.1 | - | 1 | 674 | |||
BC126156.1 | - | 5 | 1753 | 8991872 | ||
BC136298.1 | - | 5 | 1753 | 9020285 |