Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOWAHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023016.3 | NP_075392.2 | 1 | 2518 | ||
GenBank mRNA | AK023346.1 | - | 1 | 798 | ||
AK025523.1 | - | 1 | 2487 | |||
BC036652.1 | - | 1 | 2544 | 5267818 |