Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293285.1 | NP_001280214.1 | 8 | 1460 | ||
NM_001293286.1 | NP_001280215.1 | 6 | 1313 | |||
NM_023071.3 | NP_075559.2 | 7 | 1376 | |||
NR_120663.1 | 7 | 1376 | ||||
XM_011538690.2 | XP_011536992.1 | 8 | 848 | |||
XM_011538691.2 | XP_011536993.1 | 6 | 743 | |||
XM_017019840.1 | XP_016875329.1 | 7 | 816 | |||
GenBank mRNA | AB075515.1 | - | 5 | 404 | ||
AK023179.1 | - | 3 | 285 | |||
AK023202.1 | - | 7 | 775 | |||
AK095582.1 | - | 7 | 1376 | |||
AK292334.1 | - | 6 | 713 | |||
AL833614.1 | - | 8 | 901 | |||
BC048299.1 | - | 6 | 743 | 5520818 | ||
BX648916.1 | - | 6 | 685 |