Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAPRIN2 SIRNA Assays ›
Gene Symbol:
CAPRIN2
Gene Name:
caprin family member 2
Gene Aliases:
C1QDC1, EEG-1, EEG1, RNG140
Chromosome Location:
Chr.12: 30709552 - 30754994 on Build GRCh38
UniGene ID:
Hs.234355
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
130489
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAPRIN2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002259.2
NP_001002259.1
3, 4
1322
NM_001206856.2
NP_001193785.1
3, 4
1322
NM_001319842.1
NP_001306771.1
3, 4
1322
NM_001319843.1
NP_001306772.1
3, 4
1322
NM_001319844.1
NP_001306773.1
4, 5
577
NM_001319845.1
NP_001306774.1
4, 5
577
NM_001319846.1
NP_001306775.1
4, 5
567
NM_023925.4
NP_076414.2
3, 4
1322
NM_032156.4
NP_115532.3
3, 4
1322
XM_006719138.3
XP_006719201.1
3, 4
1445
XM_006719139.3
XP_006719202.1
3, 4
1445
XM_006719140.3
XP_006719203.1
3, 4
1445
XM_006719142.3
XP_006719205.1
3, 4
1445
XM_006719144.3
XP_006719207.1
3, 4
1445
XM_006719145.3
XP_006719208.1
3, 4
1445
XM_006719146.3
XP_006719209.1
3, 4
1445
XM_006719147.3
XP_006719210.1
3, 4
1445
XM_017019851.1
XP_016875340.1
3, 4
1445
XM_017019852.1
XP_016875341.1
4, 5
540
XM_017019853.1
XP_016875342.1
4, 5
610
XM_017019854.1
XP_016875343.1
3, 4
1445
XM_017019855.1
XP_016875344.1
4, 5
567
XM_017019856.1
XP_016875345.1
4, 5
567
XM_017019857.1
XP_016875346.1
4, 5
540
XM_017019858.1
XP_016875347.1
4, 5
567
XM_017019859.1
XP_016875348.1
4, 5
540
XM_017019860.1
XP_016875349.1
3, 4
1445
XM_017019861.1
XP_016875350.1
4, 5
567
XM_017019862.1
XP_016875351.1
3, 4
1445
XM_017019863.1
XP_016875352.1
3, 4
1445
XM_017019864.1
XP_016875353.1
3, 4
1445
XM_017019865.1
XP_016875354.1
3, 4
1445
XM_017019866.1
XP_016875355.1
4, 5
540
XM_017019867.1
XP_016875356.1
4, 5
567
XM_017019868.1
XP_016875357.1
4, 5
540
XM_017019869.1
XP_016875358.1
4, 5
567
XM_017019870.1
XP_016875359.1
4, 5
540
XM_017019871.1
XP_016875360.1
4, 5
611
XM_017019872.1
XP_016875361.1
4, 5
567
XM_017019873.1
XP_016875362.1
4, 5
567
XM_017019874.1
XP_016875363.1
4, 5
540
XM_017019875.1
XP_016875364.1
4, 5
567
XM_017019876.1
XP_016875365.1
2, 3
151
XM_017019877.1
XP_016875366.1
2, 3
151
XM_017019878.1
XP_016875367.1
2, 3
151
XM_017019879.1
XP_016875368.1
4, 5
540
XM_017019880.1
XP_016875369.1
2, 3
151
XM_017019881.1
XP_016875370.1
4, 5
540
XM_017019882.1
XP_016875371.1
4, 5
540
XM_017019883.1
XP_016875372.1
2, 3
151
GenBank mRNA
AK021453.1
-
3, 4
172
AY074490.1
-
3, 4
1322
AY074491.1
-
3, 4
1322
BC117672.1
-
3, 4
1119
40068103
BC117673.1
-
3, 4
1119
40068104
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of translation
negative regulation of cell growth
positive regulation of protein binding
positive regulation of peptidyl-serine phosphorylation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of dendrite morphogenesis
positive regulation of dendritic spine morphogenesis
positive regulation of canonical Wnt signaling pathway
RNA binding
receptor binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0