Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006617.1 | NP_001006618.1 | 4 | 755 | ||
NM_001006618.1 | NP_001006619.1 | 4 | 755 | |||
NM_001006619.1 | NP_001006620.1 | 4 | 755 | |||
NM_001006620.1 | NP_001006621.1 | 3 | 427 | |||
NM_001006621.1 | NP_001006622.1 | 2 | 337 | |||
NM_024117.3 | NP_077022.1 | 4 | 755 | |||
XM_011519005.1 | XP_011517307.1 | 4 | 755 | |||
XM_011519006.2 | XP_011517308.1 | 4 | 755 | |||
XM_011519007.1 | XP_011517309.1 | 4 | 755 | |||
XM_011519008.1 | XP_011517310.1 | 4 | 755 | |||
XM_011519009.1 | XP_011517311.1 | 4 | 755 | |||
XM_017015126.1 | XP_016870615.1 | 4 | 644 | |||
XM_017015127.1 | XP_016870616.1 | 4 | 644 | |||
GenBank mRNA | AK290060.1 | - | 4 | 568 | ||
AL833042.1 | - | 3 | 388 | |||
AY524429.1 | - | 4 | 752 | |||
AY524430.1 | - | 4 | 752 | |||
AY524431.1 | - | 4 | 752 | |||
AY524432.1 | - | 4 | 752 | |||
AY633624.1 | - | 4 | 722 | |||
AY633625.1 | - | 4 | 729 | |||
AY633626.1 | - | 4 | 729 | |||
BC002326.2 | - | 4 | 724 | 2823015 | ||
BC003044.1 | - | 4 | 724 | 2823015 | ||
BX438260.2 | - | 4 | 754 | |||
M37191.1 | - | 3 | 200 |