Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039885.2 | NP_001034974.1 | 4 | 2853 | ||
NM_024301.4 | NP_077277.1 | 4 | 2801 | |||
XM_005259247.2 | XP_005259304.1 | 5 | 2914 | |||
XM_005259248.2 | XP_005259305.1 | 5 | 2904 | |||
XM_005259249.4 | XP_005259306.1 | 5 | 2846 | |||
XM_011527306.1 | XP_011525608.1 | 3 | 2650 | |||
XM_011527307.1 | XP_011525609.1 | 4 | 2699 | |||
XM_017027297.1 | XP_016882786.1 | 4 | 3020 | |||
GenBank mRNA | AJ314847.1 | - | 4 | 2789 |