Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303143.1 | NP_001290072.1 | 3 | 1276 | ||
NM_024511.6 | NP_078787.2 | 2 | 1005 | |||
GenBank mRNA | AF040964.1 | - | 1 | 749 | ||
AK056134.1 | - | 2 | 1493 | |||
AK293948.1 | - | 2 | 1493 | |||
BC003648.2 | - | 3 | 1277 | 3534250 | ||
BC025356.1 | - | 1 | 517 | 4906369 | ||
CR749640.1 | - | 2 | 974 |