Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024552.2 | NP_078828.2 | 9 | 955 | ||
XM_011528290.2 | XP_011526592.1 | 9 | 991 | |||
XM_011528291.2 | XP_011526593.1 | 9 | 1108 | |||
XM_011528292.2 | XP_011526594.1 | 10 | 1931 | |||
XM_011528293.2 | XP_011526595.1 | 10 | 1107 | |||
XM_011528294.2 | XP_011526596.1 | 10 | 1075 | |||
XM_011528295.2 | XP_011526597.1 | 8 | 761 | |||
XM_017027303.1 | XP_016882792.1 | 10 | 1963 | |||
XM_017027304.1 | XP_016882793.1 | 11 | 1186 | |||
XM_017027305.1 | XP_016882794.1 | 8 | 822 | |||
GenBank mRNA | AK022151.1 | - | 9 | 915 | ||
AK055414.1 | - | 6 | 1492 | |||
AK222621.1 | - | 9 | 922 | |||
AK225394.1 | - | 9 | 918 | |||
BC009828.2 | - | 8 | 758 | 3936998 |