Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317776.1 | NP_001304705.1 | 2 | 280 | ||
NM_024672.5 | NP_078948.3 | 2 | 280 | |||
XM_005263220.2 | XP_005263277.1 | 2 | 227 | |||
XM_017008603.1 | XP_016864092.1 | 2 | 1120 | |||
XM_017008604.1 | XP_016864093.1 | 2 | 1173 | |||
XM_017008605.1 | XP_016864094.1 | 2 | 214 | |||
XM_017008606.1 | XP_016864095.1 | 2 | 262 | |||
XM_017008607.1 | XP_016864096.1 | 2 | 553 | |||
XM_017008608.1 | XP_016864097.1 | 2 | 1126 | |||
XM_017008609.1 | XP_016864098.1 | 2 | 227 | |||
XM_017008610.1 | XP_016864099.1 | 2 | 214 | |||
GenBank mRNA | AK091412.1 | - | 2 | 177 | ||
BC131722.1 | - | 2 | 161 | |||
BX648134.1 | - | 2 | 385 | |||
DA317935.1 | - | 2 | 246 | |||
DB093127.1 | - | 2 | 281 |