Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024786.2 | NP_079062.1 | 1 | 590 | ||
XM_017009869.1 | XP_016865358.1 | 2 | 306 | |||
XM_017009870.1 | XP_016865359.1 | 4 | 2117 | |||
XM_017009871.1 | XP_016865360.1 | 4 | 2066 | |||
XM_017009872.1 | XP_016865361.1 | 2 | 306 | |||
GenBank mRNA | AK023215.1 | - | 1 | 590 | ||
BC032000.1 | - | 1 | 209 | 4750082 |