Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024798.2 | NP_079074.2 | 6 | 489 | ||
NR_073534.1 | 6 | 536 | ||||
XM_005254677.3 | XP_005254734.1 | 5 | 424 | |||
XM_017022581.1 | XP_016878070.1 | 5 | 415 | |||
GenBank mRNA | AK024014.1 | - | 6 | 489 | ||
AK126655.1 | - | 5 | 407 | |||
AL832838.1 | - | 2 | 243 | |||
AY044653.1 | - | 6 | 430 | |||
BC019655.1 | - | 6 | 514 | 4932954 | ||
BC030225.1 | - | 4 | 1165 |