Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TULP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160408.1 | NP_001153880.1 | 2, 3 | 184 | ||
NM_003324.4 | NP_003315.2 | 2, 3 | 184 | |||
GenBank mRNA | AF045583.1 | - | 2, 3 | 148 | ||
AK024246.1 | - | 2, 3 | 184 | |||
BC032587.1 | - | 2, 3 | 157 | 5501868 |