Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0226L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286761.1 | NP_001273690.1 | 15 | 2758 | ||
NM_001286762.1 | NP_001273691.1 | 13 | 2510 | |||
NM_001286763.1 | NP_001273692.1 | 15 | 2393 | |||
NM_001286764.1 | NP_001273693.1 | 14 | 2277 | |||
NM_001286765.1 | NP_001273694.1 | 14 | 2123 | |||
NM_001286766.1 | NP_001273695.1 | 13 | 2007 | |||
NM_025113.3 | NP_079389.2 | 15 | 2780 | |||
GenBank mRNA | AK025215.1 | - | 13 | 2259 | ||
AK125750.1 | - | 5 | 1501 | |||
AK125950.1 | - | 15 | 2765 | |||
AK300183.1 | - | 14 | 1827 | |||
AK307854.1 | - | 13 | 1749 | |||
BC004495.1 | - | 11 | 1477 | |||
BC021097.1 | - | 6 | 1130 | |||
BC032311.1 | - | 13 | 3807 | |||
BC043488.1 | - | 15 | 2126 | 5205680 | ||
BX641126.1 | - | 13 | 1814 |