Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270367.1 | NP_001257296.1 | 4 | 412 | ||
NM_001270368.1 | NP_001257297.1 | 4 | 425 | |||
NM_001270369.1 | NP_001257298.1 | 5 | 464 | |||
NM_001270370.1 | NP_001257299.1 | 6 | 471 | |||
NM_001270371.1 | NP_001257300.1 | 5 | 486 | |||
NM_001270372.1 | NP_001257301.1 | 4 | 404 | |||
NM_001270373.1 | NP_001257302.1 | 5 | 443 | |||
NM_001270374.1 | NP_001257303.1 | 5 | 793 | |||
NM_025125.3 | NP_079401.2 | 4 | 391 | |||
NR_072984.1 | 5 | 408 | ||||
NR_072985.1 | 5 | 415 | ||||
NR_072986.1 | 4 | 313 | ||||
NR_072987.1 | 5 | 365 | ||||
XM_006717978.2 | XP_006718041.1 | 5 | 743 | |||
XM_017016679.1 | XP_016872168.1 | 6 | 583 | |||
GenBank mRNA | AK023325.1 | - | 4 | 367 | ||
AK222536.1 | - | 4 | 372 | |||
AK300490.1 | - | 4 | 422 | |||
AK309425.1 | - | 5 | 690 | |||
AK309980.1 | - | 4 | 400 | |||
AK310160.1 | - | 5 | 373 | |||
AK311563.1 | - | 5 | 381 | |||
AL832397.1 | - | 4 | 288 | |||
AL832483.1 | - | 4 | 399 | |||
BC022252.1 | - | 4 | 367 | 4668550 | ||
BP204746.1 | - | 6 | 440 | |||
BP293793.1 | - | 5 | 418 | |||
BX537803.1 | - | 5 | 373 | |||
CR457310.1 | - | 4 | 337 | |||
CX756014.1 | - | 6 | 427 | |||
DA263618.1 | - | 5 | 323 | |||
DA573956.1 | - | 5 | 424 | |||
DA880596.1 | - | 5 | 735 | |||
DA930892.1 | - | 4 | 380 | |||
DB485367.1 | - | 5 | 415 |